Learn More
Invitrogen™ Platinum™ SuperFi™ U Multiplex Master Mix
Invitrogen Platinum SuperFi U Multiplex Master Mix is a product designed to synthesize multiple custom amplicons for further use in next-generation sequencing.
Brand: Invitrogen™ A5140024
21.97 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 0.00600kg
Description
- Designed to synthesize multiple custom amplicons for further use in next-generation sequencing
- The ready-to-use 4X master mix contains uracil-tolerant high-fidelity DNA polymerase with all necessary buffer components and dNTPs for efficient targeted amplification
- Optimized for high multiplex PCR
- Enables fast, efficient, and robust amplification
- Provides high yields
Specifications
Transparent | |
Store at -25°C to -15°C | |
Sars-Covid-2 Sequencing | |
24 preps | |
Master Mix |
4X | |
Platinum™ SuperFi™ U DNA Polymerase | |
24 Preps | |
Collibri™ | |
All |