Learn More
Emma Safety Footwear Pluvius Safety Shoes
Protected with Sympatex™ one of the hardest wearing, most liquid repellant materials available, including corrosive chemicals. Provides outstanding grip in every kind of weather and a good on-foot fit.
Brand:  Emma Safety Footwear MM93786842
 View more versions of this product
Product Code. 13439599
  • £233.00 / 1 pair
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • S3-Safety shoe with sole plate for wet work
  • Full grain leather / Sympatex™ construction
  • Chemical resistant / Liquid repellant
  • GripForce™ Duo Heat Resistant Outsole
  • Sanitized
Specifications
Specifications
EN ISO 20345:2011
Black, Gray
Hydro-Tec™ Sanitized Silver / Sympatex™
42
Chemicals
Laces
Pluvius
Leather
Steel