Learn More
Emma Safety Footwear Pluvius Safety Shoes
Protected with Sympatex™ one of the hardest wearing, most liquid repellant materials available, including corrosive chemicals. Provides outstanding grip in every kind of weather and a good on-foot fit.
Brand: Emma Safety Footwear MM93786846
This item is not returnable.
View return policy
Description
- S3-Safety shoe with sole plate for wet work
- Full grain leather / Sympatex™ construction
- Chemical resistant / Liquid repellant
- GripForce™ Duo Heat Resistant Outsole
- Sanitized
Specifications
EN ISO 20345:2011 | |
Black, Gray | |
Hydro-Tec™ Sanitized Silver / Sympatex™ | |
46 | |
Chemicals |
Laces | |
Pluvius | |
Leather | |
Steel |