Learn More
PLVAP Mouse anti-Human, Unconjugated, Clone: 1D4A6, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67158-1-IG-150UL
 View more versions of this product
Product Code. 16819635
  • £349.00 / 150µL
Explore more special offers
Description
Description
The PLVAP antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PLVAP. This antibody recognizes human, pig antigen. The PLVAP antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
PLVAP
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Pig
Western Blot, ELISA
1D4A6
Unconjugated
150 μL
83483
IgG2a