missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLVAP Mouse anti-Human, Unconjugated, Clone: 1D4A6, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
Antigen | PLVAP |
---|---|
Clone | 1D4A6 |
Concentration | 1000 μg/mL |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16829635
|
Proteintech
67158-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16819635
|
Proteintech
67158-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The PLVAP antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PLVAP. This antibody recognizes human, pig antigen. The PLVAP antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
PLVAP | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
83483 | |
Primary |
1D4A6 | |
Western Blot, ELISA | |
Unconjugated | |
Human, Pig | |
IgG2a |