missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PolarScreen™ ER Alpha Competitor Assay, Green

Brand:  Thermo Scientific™ A15882

 View more versions of this product

Product Code. 15340186

  • £2095.00 / Each
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The PolarScreen™ ER (Estrogen Receptor) Alpha Competitor Assay Kit, Green, provides a sensitive and efficient method for high-throughput, fluorescence polarization-based screening of potential ER alpha ligands. The kit uses insect cell-expressed, full-length, native (untagged) human estrogen receptor alpha (available separately, Cat. No. A15674) and a novel, high-affinity, fluorescent estrogen ligand (Fluormone ES2 Green) in a homogenous, mix-and-read assay format.

This kit produces a green color read-out; a 'red' kit is also available. Both kits suitably determine the IC50s of competitive compounds. Color choice depends on the spectral properties of the compounds to be screened and the available instrumentation. For additional information or assistance contact drugdiscoverytech@lifetech.com.

PolarScreen™ ER Alpha Competitor Assay, Green, is:

  • Complete & ready-to-use out of the box—all reagents included, just add your test compounds
  • High throughput compatible—800 x 20μL fluorescence polarization assays in a 384-well plate
  • Convenient—available in bulk sizes for larger screening needs

    How the Assay Works
    Full length ER alpha is added to a fluorescent estrogen ligand to form an ER-Fluormone ES2 complex. This complex is added to individual test compounds in multiwell plates. To determine relative affinity, the assay measures fluorescence polarization. Competing test compounds will displace the Fluormone ES2 ligand from ER alpha, permitting it to tumble rapidly and resulting in low polarization values. A shift in the fluorescence polarization value in the presence of test compound is used to determine the relative affinity of test compounds for ER alpha.

    All components are manufactured under the strictest quality parameters. This product's performance, per the Certificate of Analysis, is guaranteed for 6 months after purchase. Contact us for bulk quantities or for assistance with other nuclear receptor assay needs, from assay development to library screening and beyond.
    TRUSTED_SUSTAINABILITY
  • Specifications

    Specifications

    Contains:

    • 4 tubes full-length ER Alpha Recombinant Protein (store at -68 to -85°C)
    • 1 tube of Fluormone ES2 Green Tracer (store at -5 to -30°C)
    • 2 bottles of ES2 Screening Buffer (store at -5 to -30°C)
    Fluorescent Imager
    2500 Assays
    Fluorescence
    384-well Plate
    Competitor Assay Kit
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

    For Research Use Only. Not for use in diagnostic procedures.