missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PolarScreen™ Progesterone Receptor Competitor Assay Kit, Red

The PolarScreen™ Progesterone Receptor Competitor Assay Kit, Red, provides a sensitive and efficient method for high-throughput, fluorescence polarization-based screening of potential progesterone receptor (PR) ligands.

Brand:  Thermo Scientific™ A15906

Product Code. 15667858

  • £900.00 / Pack of 400
Estimated Shipment: 07-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The PolarScreen™ Progesterone Receptor Competitor Assay Kit, Red, provides a sensitive and efficient method for high-throughput, fluorescence polarization-based screening of potential progesterone receptor (PR) ligands. The kit uses insect cell-expressed, human progesterone receptor ligand binding domain tagged with glutathione-S-transferase (GST) (also available separately) and a novel, high-affinity, fluorescent progesterone ligand (Fluormone™ PL Red) in a homogenous, mix-and-read assay format.

This kit produces a red color read-out; a 'green' kit is also available. Both kits suitably determine the IC50s of competitive compounds. Color choice depends on the spectral properties of the compounds to be screened and the available instrumentation. For additional information or assistance contact us at drugdiscoverytech@lifetech.com.

The PolarScreen Progesterone Receptor Competitor Assay is:

  • Complete & ready-to-use out of the box—all reagents included, just add your test compounds
  • High throughput compatible—fluorescence polarization assays in a 384-well plate
  • Convenient—components available in bulk sizes for larger screening needs

How the Assay Works

An N-terminal fusion of glutathione transferase to the ligand-binding domain of the human progesterone receptor (PR-LBD(GST)) is added to a fluorescently-tagged progesterone ligand (Fluormone PL Red) in the presence of competitor test compounds in microwell plates. The presence of effective competitors prevents the formation of a PL Red/PR-LBD(GST) complex resulting in a decrease of the polarization value. The shift in polarization value in the presence of test compounds is used to determine relative affinity of test compounds for PR-LBD(GST).

Contact us for bulk quantities or for assistance with other nuclear receptor assay needs, from assay development to library screening and beyond.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Competitive Binding Assay, Nuclear Receptor Assay
• Progesterone Receptor Ligand Binding Domain Recombinant Protein, 1 tube (store at -68°C to -85°C)
• Fluormone PL Red Tracer, 1 tube (-5°C to -30°C)
• PR Red Screening Buffer, 2 bottles (-5°C to -30°C)
• DTT, 1 tube (-5°C to -30°C)
PR
400 x 20 μL Assays
400 x 20 μL assays
PolarScreen™
Assay Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.