missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poly(propylene glycol), average M.W. 2.000, Thermo Scientific Chemicals
£12.40 - £160.00
Chemical Identifiers
CAS | 25322-69-4 |
---|---|
Molecular Formula | (C3H6O)n |
Molecular Weight (g/mol) | 134.18 |
MDL Number | MFCD00084446 |
InChI Key | DNIAPMSPPWPWGF-UHFFFAOYNA-N |
Synonym | polypropylene glycol, polyoxypropylene, 2-2-hydroxypropoxy propan-1-ol, emkapyl, lineartop e, niax ppg, polyoxypropylene glycol, methyloxirane homopolymer, desmophen 360c, 2-2-hydroxypropoxy-1-propanol |
PubChem CID | 32881 |
IUPAC Name | 2-(2-hydroxypropoxy)propan-1-ol |
SMILES | [H]OC(C)CO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10489890
|
Thermo Scientific Acros
192180050 |
5 g | Glass bottle | ||||||
10740701
|
Thermo Scientific Acros
192185000 |
500 g | Glass bottle |
£52.30
500g |
In Stock
Log in to see stock. |
||||
10256963
|
Thermo Scientific Acros
192180025 |
2.5 kg | Glass bottle |
£160.00
2.50kg |
In Stock
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
25322-69-4 | |
134.18 | |
DNIAPMSPPWPWGF-UHFFFAOYNA-N | |
32881 | |
[H]OC(C)CO |
(C3H6O)n | |
MFCD00084446 | |
polypropylene glycol, polyoxypropylene, 2-2-hydroxypropoxy propan-1-ol, emkapyl, lineartop e, niax ppg, polyoxypropylene glycol, methyloxirane homopolymer, desmophen 360c, 2-2-hydroxypropoxy-1-propanol | |
2-(2-hydroxypropoxy)propan-1-ol |
Specifications
White to Gray | |
Crystals or Crystalline Powder | |
(C3H6O)n | |
MFCD00084446 | |
polypropylene glycol, polyoxypropylene, 2-2-hydroxypropoxy propan-1-ol, emkapyl, lineartop e, niax ppg, polyoxypropylene glycol, methyloxirane homopolymer, desmophen 360c, 2-2-hydroxypropoxy-1-propanol | |
[H]OC(C)CO | |
134.18 | |
53 to 58 mg KOH/g | |
300 cSt (25°C) | |
Poly(propylene glycol), Average M.W. 2.000 |
25322-69-4 | |
0.1 mg KOH/g max. | |
H[OCH(CH3)CH2]nOH | |
1.0000g/mL | |
DNIAPMSPPWPWGF-UHFFFAOYNA-N | |
2-(2-hydroxypropoxy)propan-1-ol | |
32881 | |
Authentic | |
0.15% Max. |
Safety and Handling
missing translation for 'rtecsNumber' : OE5784360
missing translation for 'tsca' : TSCA
RUO – Research Use Only