missing translation for 'onlineSavingsMsg'
Learn More

Poly(styrene-divinylbenzene), aminomethylated, 1% cross-linked, 200-400 mesh, Thermo Scientific™

£75.20 - £760.00

Chemical Identifiers

CAS 89551-24-6
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.156
MDL Number MFCD00146442
InChI Key WGQKYBSKWIADBV-UHFFFAOYSA-N
Synonym benzylamine, benzenemethanamine, monobenzylamine, alpha-aminotoluene, phenylmethyl amine, aminomethyl benzene, 1-phenylmethanamine, moringine, n-benzylamine, phenylmethylamine
PubChem CID 7504
ChEBI CHEBI:40538
IUPAC Name phenylmethanamine
SMILES C1=CC=C(C=C1)CN
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11428590
View Documents
Thermo Scientific Alfa Aesar
044394.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11408590
View Documents
Thermo Scientific Alfa Aesar
044394.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11418590
View Documents
Thermo Scientific Alfa Aesar
044394.22
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

89551-24-6
107.156
WGQKYBSKWIADBV-UHFFFAOYSA-N
7504
phenylmethanamine
C7H9N
MFCD00146442
benzylamine, benzenemethanamine, monobenzylamine, alpha-aminotoluene, phenylmethyl amine, aminomethyl benzene, 1-phenylmethanamine, moringine, n-benzylamine, phenylmethylamine
CHEBI:40538
C1=CC=C(C=C1)CN

Specifications

89551-24-6
C7H9N
benzylamine, benzenemethanamine, monobenzylamine, alpha-aminotoluene, phenylmethyl amine, aminomethyl benzene, 1-phenylmethanamine, moringine, n-benzylamine, phenylmethylamine
C1=CC=C(C=C1)CN
107.156
CHEBI:40538
Poly (styrene-divinylbenzene), Aminomethylated, 1% cross-linked
Powder
MFCD00146442
WGQKYBSKWIADBV-UHFFFAOYSA-N
phenylmethanamine
7504
200 to 400 Mesh
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications