missing translation for 'onlineSavingsMsg'
Learn More

Quickfit™ Polyacetal Joint Clips

Manufactured from polyacetal for strength and rigidity. Quickfit™ Polyacetal Clamps for Conical Joints are designed to secure corresponding Quickfit™ glassware with conical joints.

Brand:  Quickfit™ KC12

45.72 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 10796461

  • £60.30 / Pack of 10
Estimated Shipment: 03-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Manufactured from polyacetal for strength and rigidity. Quickfit™ Polyacetal Clamps for Conical Joints are designed to secure corresponding Quickfit™ glassware with conical joints.

  • Effortless attachment and removal of clips
  • Reduces the need for traditional clamping
  • Withstands temperatures up to 80°C
  • Color coded sizes for quick identification
TRUSTED_SUSTAINABILITY
Specifications

Specifications

10/Pk.
Polyacetal
Joint Size 12/21
Joint Clamp
Glassware with conical joints
Violet
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.