missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Polyester Photolum Marking Markers

Brand:  Brady™ 224076

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11741965

  • £12.35 / Each
Estimated Shipment: 06-09-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

ASTM E–2072-00 / ASTM E–2073-00; ASTM E-162; ASTM E-648; ASTM E-662; IMO Resolution A.752 (18); ISO / CD 15370; Marine Safety Committee MSC.27 (61); DIN 67 510 (parts 1-4) and PSPA Standard 002 Part 2. Meets OSHA 1910.37; Lloyd's Register SAS F050294.p
1 Ea.
Polyester long afterglow
255 mm
100 mm
100 x 255 mm
Door Sign
Polyester
Permanent Acrylic
Exit and directional signs, identification of fire alarms and extinguishers, low location egress pathway markings, and directional systems for escape or evacuations routes
0.049 mm
-40°C to 80°C
Door handle background - Right turn
Photoluminescent on Green
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers