missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Polyester Safety Tags
Polyester Safety Tags provide on-the-spot identification of safety measures. The tag is printed on both sides, making sure the vital information is always visible.
£26.75 - £28.45
Specifications
Product Type | Safety Tag |
---|---|
Material | Polyester |
Certifications/Compliance | OSHA 1910.145 |
Durability | 3 to 5 years |
For Use With (Application) | Safety Identification |
Product Code | Brand | Displayed Text | Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Displayed Text | Color | Price | Quantity & Availability | ||||
12788029
|
Brady™
241703 |
Caution Men working on this equipment | Black/Yellow on White |
£26.75
1 pack |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||||
12778029
|
Brady™
241713 |
Caution Biohazard | Black/Yellow on White |
£26.75
1 pack |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||||
12798029
|
Brady™
241701 |
Caution Out of Order | Black/Yellow on White |
£28.45
1 pack |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
12713385
|
Brady™
241708 |
Danger Electric shock risk | Black/Yellow on White |
£26.75
1 pack |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
12787445
|
Brady™
241710 |
Danger Live wire | Black/Yellow on White |
£26.75
1 pack |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||||
12757455
|
Brady™
241714 |
Danger Radioactive material | Black/Yellow on White |
£26.75
1 pack |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||||
12707465
|
Brady™
241700 |
Danger | Black/Yellow on White |
£28.45
1 pack |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||||
12787485
|
Brady™
241718 |
Do not switch on under Maintenance | Black/Red on White |
£26.75
1 pack |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||||
12777485
|
Brady™
241717 |
Do not switch on | Black/Red on White |
£26.75
1 pack |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||||
12797485
|
Brady™
241719 |
Do not Touch | Black/Red on White |
£26.75
1 pack |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
12727495
|
Brady™
241716 |
Do not use Out of Order | Black/Red on White |
£28.45
1 pack |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||||
12737495
|
Brady™
241720 |
Failed Electrical Safety test | Black/Red on White |
£26.75
1 pack |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||||
12747505
|
Brady™
241721 |
Tested For Electrical Safety | Black/Green on White |
£26.75
1 pack |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||||
Description
- The tag is printed on both sides, making sure the vital information is always visible.
Specifications
Safety Tag | |
OSHA 1910.145 | |
Safety Identification | |
145 mm | |
10 Pk. | |
-40°C to +100°C | |
3.35 in. |
Polyester | |
3 to 5 years | |
5.71 in. | |
(1) 15 x 140 mm Attachment Tie | |
-40°F to +212°F | |
0.249 mm | |
85 mm |