missing translation for 'onlineSavingsMsg'
Learn More

Cytiva PEEK Sample Loops

Resists organic solvents and salt buffers commonly used in chromatography of biomolecules. Cytiva Life Sciences™ PEEK Sample Loops are an important accessory for chromatography applications.

Brand:  Cytiva 18-1113-98

 View more versions of this product

Product Code. 11360032

  • £171.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Wetted parts are resistant to organic solvents
  • Wetted parts are resistant to salt buffers
  • Except 100 % Ethyl acetate, 100 % Hexane and 100 % Tetrahydrofuran
  • Polyetheretherketone (PEEK)
 
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Sample Loop
1.58 mm
100 μL
PEEK
0.5 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers