Learn More
Polyethylene glycol 8,000, Thermo Scientific Chemicals
Description
Polyethylene glycol is used in the modification of therapeutic proteins and peptides to enhance its solubility. It is also used in the fabrication of bioactive and immuno isolating barriers for encapsulation of cells. In gas chromatography, it is used as a polar stationary phase. It is used as mold release agent and lubricant in fabricating elastomers. As a dispersant, it is used in toothpastes. It finds application in blood banking as a potentiator, which is used to detect antigens and antibodies. In pharmaceutical industry, it is used as tablet and capsule lubricant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | White |
pH | 5 to 7 |
Physical Form | Powder |
Melting Point | 55°C to 63°C |
Merck Index | 14,7568 |
Solubility Information | Soluble in water and aromatic hydrocarbons. Slightly soluble in aliphatic hydrocarbons and organic solvents. |
Quantity | 500 g |
Odor | Odorless |
Chemical Name or Material | Polyethylene glycol 8,000 |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
N12K003 | Certificate of Analysis | 11/03/2025 | 043443.A9,043443.22,043443.A3,043443.36 |
U12K036 | Certificate of Analysis | 13/01/2025 | 043443.A9,043443.22,043443.A3,043443.36 |
U08K036 | Certificate of Analysis | 29/08/2024 | 043443.A9,043443.22,043443.A3,043443.36 |
U08K022 | Certificate of Analysis | 29/08/2024 | 043443.A9,043443.22,043443.A3,043443.36 |
U08K023 | Certificate of Analysis | 29/08/2024 | 043443.A9,043443.22,043443.A3,043443.36 |
Special Offers
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
New Offer Save Up to 70%
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.