missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene glycol monomethyl ether methanesulfonate, M.W. 5,000, Thermo Scientific™

£16.60 - £209.00

Chemical Identifiers

CAS 175172-61-9
Molecular Formula C4H10O4S
Molecular Weight (g/mol) 154.18
MDL Number MFCD03100948
InChI Key BCKAHDGFNHDQST-UHFFFAOYSA-N
Synonym methanesulfonic acid 2-methoxyethyl ester, ethanol, 2-methoxy-, methanesulfonate, 2-methoxyethanol methanesulfonate, 2-methoxyethyl mesylate, 2-methoxyethylmethanesulfonate, acmc-1c990, 2-methoxyethyl methane sulfonate, methanesulfonic acid2-methoxyethyl ester, ethanol, 2-methoxy-, 1-methanesulfonate, methanesulfonic acid 2-methoxy-ethyl ester
PubChem CID 146293
IUPAC Name 2-methoxyethyl methanesulfonate
SMILES COCCOS(=O)(=O)C
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15407225
View Documents
Thermo Scientific Alfa Aesar
046454.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15487215
View Documents
Thermo Scientific Alfa Aesar
046454.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15497215
View Documents
Thermo Scientific Alfa Aesar
046454.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

175172-61-9
154.18
BCKAHDGFNHDQST-UHFFFAOYSA-N
146293
COCCOS(=O)(=O)C
C4H10O4S
MFCD03100948
methanesulfonic acid 2-methoxyethyl ester, ethanol, 2-methoxy-, methanesulfonate, 2-methoxyethanol methanesulfonate, 2-methoxyethyl mesylate, 2-methoxyethylmethanesulfonate, acmc-1c990, 2-methoxyethyl methane sulfonate, methanesulfonic acid2-methoxyethyl ester, ethanol, 2-methoxy-, 1-methanesulfonate, methanesulfonic acid 2-methoxy-ethyl ester
2-methoxyethyl methanesulfonate

Specifications

White
Granular Solid
C4H10O4S
methanesulfonic acid 2-methoxyethyl ester, ethanol, 2-methoxy-, methanesulfonate, 2-methoxyethanol methanesulfonate, 2-methoxyethyl mesylate, 2-methoxyethylmethanesulfonate, acmc-1c990, 2-methoxyethyl methane sulfonate, methanesulfonic acid2-methoxyethyl ester, ethanol, 2-methoxy-, 1-methanesulfonate, methanesulfonic acid 2-methoxy-ethyl ester
COCCOS(=O)(=O)C
154.18
Polyethylene glycol monomethyl ether methanesulfonate, M.W. 5,000
175172-61-9
58°C to 61°C
MFCD03100948
BCKAHDGFNHDQST-UHFFFAOYSA-N
2-methoxyethyl methanesulfonate
146293
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.