Learn More
Masterflex™ Polyethylene Male Pipe Adapter
Designed for quick and easy assembly without tools
Brand: Masterflex™ 30621-54
Code : A0
Additional Details : Weight : 0.80000kg
Description
- Polyethylene Male Pipe Adapter
- Adapts from a NPT male thread to a hose barb
- Barbed fittings are designed for quick and easy assembly without tools
- Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30psi)
- Available in packs of ten
Specifications
Male Pipe Adapter | |
Not Autoclavable | |
1/2 in. | |
1/2 in. | |
1/4 x 1/2 in. |
Barb x NPT(M) Connection | |
HDPE | |
1/4 in. | |
Ethylene Oxide |