Learn More
Masterflex™ Polyethylene Male Pipe Adapter
Designed for quick and easy assembly without tools
Brand:  Masterflex™ 30621-12
 View more versions of this product
Product Code. 11765259
  • £26.50 / Pack of 10
Estimated Shipment: 03-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Polyethylene Male Pipe Adapter
  • Adapts from a NPT male thread to a hose barb
  • Barbed fittings are designed for quick and easy assembly without tools
  • Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30psi)
  • Available in packs of ten
Specifications
Specifications
Barb x NPT(M) Connection
HDPE
3/4 in.
3/4 in.
3/4 x 3/4 in.
Male Pipe Adapter
Not Autoclavable
10 Fittings
3/4 in.
Ethylene Oxide
Straight
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Masterflex™ Polyethylene Male Pipe Adapter > 3/4 x 3/4 in.