Learn More
Masterflex™ Polyethylene Male Pipe Adapter
Designed for quick and easy assembly without tools
Brand: Masterflex™ 30621-70
This item is not returnable.
View return policy
Description
- Polyethylene Male Pipe Adapter
- Adapts from a NPT male thread to a hose barb
- Barbed fittings are designed for quick and easy assembly without tools
- Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30psi)
- Available in packs of ten
Specifications
Barb x NPT(M) Connection | |
HDPE | |
3/4 in. | |
3/4 in. | |
1/2 x 3/4 in. | |
Male Pipe Adapter |
Not Autoclavable | |
10 Fittings | |
1/2 in. | |
Ethylene Oxide | |
Straight |
Product Content Correction
Product Title
Masterflex™ Polyethylene Male Pipe Adapter > 1/2 x 3/4 in.
Spot an opportunity for improvement?Share a Content Correction