missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ SIL G-25 / UV254 Glass Plates

Stable in almost all organic solvents. Macherey-Nagel™ SIL G-25 / UV254 Glass Plates are analytical plates with unmodified standard silica layers on glass for use in TLC. Feature a green fluorescence indicator for short-wave UV.

Brand:  Macherey-Nagel™ 809028.100

134.73 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 12810951

  • £204.88 / Pack of 200
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

SIL G-25 / UV254 Glass Plates feature unmodified, standard-grade silica 60 layers with a green fluorescent indicator.

  • Manganese-activated zinc silicate indicator with green fluorescence for short-wave UV (254nm)
  • Highly polymeric products, which are stable in almost all organic solvents and resistant towards aggressive visualization reagents
Specifications

Specifications

Glass Plates with Unmodified Silica 60 Layers
60 Å
Glass TLC Plate
0.75 mL/g
0.25 mm
5 to 17 μm
100 Pk.
UV254
∼500 m2/g
2.5 x 7.5 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Macherey-Nagel™ SIL G-25 / UV254 Glass Plates > Plate Size: 25 x 75mm; Quantity: 100 Plates

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.