Learn More
Scilabub™ Polyimide Cryogenic Glove
Polyimide Cryogenic Gloves have been specially developed for the handling of very cold objects stored in the vapour phase of liquid nitrogen (ranging from -120°C to -190°C).
Brand:  Scilabub™ GLO/CMEL
 View more versions of this product
Product Code. 12981185
  • £354.00 / 1 pair
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Developed for handling very cold objects, typically those stored in the vapour phase of liquid nitrogen (in the range -120°C to -190°C)
  • Gloves should not actually be immersed in liquid nitrogen
  • The outer layer is waterproof material to resist condensed water on cold surfaces
  • Permeable to gases (including water vapour) for enhanced user comfort so that there is no clammy condensation inside
  • Stitch holes unsealed to reduce bulk around the fingers
  • Very durable, and retain flexibility (albeit with some stiffening) for long periods in cold atmospheres
  • Polyimide outer material, polyolefin with some polyester as filling and cotton liner
  • Manufactured to EN511
Specifications
Specifications
PPE Category III
Blue
Powder Free
Large
-190°C
1 Pack
500 mm
Polyimide
Fabric
Cryogenic Gloves
Non-sterile
Cotton
Water