missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ Polyimide Tubing

Use as an electrical and thermal insulator, for fluid and gas delivery, or as a tough sheath for glass fibers

Brand:  Masterflex™ 95820-07

 View more versions of this product

Product Code. 15173523

  • £130.00 / Each
Estimated Shipment: 11-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Excellent chemical, heat, radiation and abrasion resistance.
  • High dielectric strength.
  • Clear amber color.
  • Resin meets USP Class VI requirements and NEMA standards.
  • Temperature range: -418 to +896°F (-250 to +480°C).
  • Sterilize: Autoclave, ethylene oxide or gamma irradiation.
Specifications

Specifications

ADCF;European Pharmacopoeia;NSF-51;NSF-61;Non-Phthalate;USP;3A;FDA;Non-DEHP
0.76 mm
0.965 mm
1.5 m
Use as an electrical and thermal insulator, for fluid and gas delivery, or as a tough sheath for glass fibers.
-250°C
Excellent chemical, heat, radiation, and abrasion resistance; high dielectric strength.
Clear Amber
896°F
Autoclavable
Polyamide
5 ft.
0.0508 mm
-418°F
5 ft./Pk.
Autoclave, ethylene oxide, or gamma irradiation.
550 psi
480°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.