Learn More
Lenz Laborglasinstrumente™ Keck™ POM Conical Joint Clips
Ensures a secure connection while preventing damage to glass tubing and increasing safety in the laboratory. Lenz Laborglasinstrumente™ Keck™ POM Conical Joint Clips are manufactured from temperature resistant polyoxymethylene (POM).
Brand: Lenz Laborglasinstrumente™ 01103710
Additional Details : Weight : 0.60000kg
Description
- Good spring properties
- Color coded by size
- Will not corrode or bend out of shape
Specifications
10/Pk. | |
POM | |
Turquoise |
Joint Clip | |
For conical ground joints |