Learn More
Alpha Packaging™ Polypropylene Sample Bag
Use a Alpha Packaging™ Polypropylene Sample Bag to package and heat seal samples. The bag is available in two sizes.
Brand: Alpha Packaging™ 12390499
Description
- Heat seal closure
- Clear
- Polypropylene
Specifications
Bag | |
Polypropylene | |
250 mil |
Heat Seal | |
250 Pack | |
450 x 600mm |