missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpha Packaging™ Polypropylene Sample Bag
Use a Alpha Packaging™ Polypropylene Sample Bag to package and heat seal samples. The bag is available in two sizes.
£280.00 - £491.00
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | ||||||
12360499
|
Alpha Packaging™
12360499 |
150 x 200mm |
£491.00
Pack of 500 |
Estimated Shipment: 01-04-2025
Log in to see stock. |
||||||
12390499
|
Alpha Packaging™
12390499 |
450 x 600mm |
£280.00
Pack of 250 |
In Stock
Log in to see stock. ![]() |
||||||
Description
- Heat seal closure
- Clear
- Polypropylene
Specifications
Bag | |
Polypropylene | |
250 mil |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Alpha Packaging™ Polypropylene Sample Bag