Learn More
Cole-Parmer™ Polypropylene Cable Ties
Keep items bundled during autoclaving with these ties. Cole-Parmer™ Polypropylene Cable Ties are ideal for tubing bundles.
Brand: Cole-Parmer™ 06830-95
Code : A0
Additional Details : Weight : 0.80000kg
Description
Keep items bundled during autoclaving with these ties. Cole-Parmer™ Polypropylene Cable Ties are ideal for tubing bundles.
- Polypropylene (PP) cable ties resist a number of chemicals
- UV-resistant
- Sold in packs of 1000
Specifications
Cable Tie | |
Black | |
1000/Pk. |
Polypropylene | |
20 cm | |
Ideal for tubing bundles |