Learn More
Buerkle™ Polypropylene Reducing Hose Connectors
Use Buerkle™ Polypropylene Reducing Hose Connectors to make a connections between larger and smaller hoses.
£24.20 - £37.30
Specifications
Product Type Reducing Hose Connector
Certifications/Compliance EU Foodstuffs, RoHS
Autoclavable Autoclavable
Material Polypropylene
Packaging Quantity 10/Pk.; 10/Scale
View More Specs
Products 3
Product Code Brand For Use With Price Quantity & Availability  
Product Code Brand For Use With Price Quantity & Availability  
10642482
View Documents Promotion Details
Buerkle™
8703-0408
4 to 8/8 to 12 mm Hose
£30.80
Pack of 10
In Stock
to see stock.
Edge
Add to basket
10630143
View Documents Promotion Details
Buerkle™
8703-0412
4 to 8/12 to 16 mm Hose
£24.20
Pack of 10
In Stock
to see stock.
Add to basket
10739792
View Documents Promotion Details
Buerkle™
8703-0812
8 to 12/12 to 16 mm Hose
£37.30
Pack of 10
In Stock
to see stock.
Edge
Add to basket
Description
Description
  • Polypropylene
  • Sterilizable
Specifications
Specifications
Reducing Hose Connector
Autoclavable
10/Pk.; 10/Scale
EU Foodstuffs, RoHS
Polypropylene
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Buerkle™ Polypropylene Reducing Hose Connectors