missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Polypropylene Floating Microtube Racks

Keep microtubes afloat in a water bath or push into crushed ice for quick cooling. Fisherbrand™ Floating Microtube Racks can be used on a benchtop, with water baths, or in ice containers.

Brand:  Fisherbrand™ HS2132A

43.38 GBP valid until 2025-07-11
Use promo code "25721" to get your promotional price.


 View more versions of this product

Product Code. 13436959

  • £49.87 / Pack of 4
Estimated Shipment: 29-04-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Detachable legs provide two options:
    • Attach legs so rack stands upright on bench top
    • Detach legs so samples can be quickly popped out by pushing down onto any surface.
  • Measure 60mm high including legs
  • Not recommended for autoclaving
Specifications

Specifications

Not Autoclavable
16 1.5/2.0 mL Microtubes
Polypropylene
102 mm
60 mm
White
Detachable legs
Square
102 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Polypropylene Floating Microtube Racks > No. of Positions: 16 Wells

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.