missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Polypropylene Funnel with Fluted Interior

Polypropylene construction. Short wide stem provides smooth, quick funneling.

Brand:  BRAND™

10.25 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 10588901

  • £19.45 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Polypropylene construction
  • Short, wide stem
  • Includes internal fluting
TRUSTED_SUSTAINABILITY
Specifications

Specifications

150 mm
14 mm
Polypropylene
115 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BRAND™ Polypropylene Funnel with Fluted Interior > Stem Length: 115 mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.