Learn More
Brady™ Polypropylene Gate Valve Lockout
This Polypropylene Gate Valve Lockout has a unique knockout center that accommodates OS and Y or rising stem valves. Lockout accepts locks with shackles up to 9.53 mm diameter. Danger labels in English, Spanish and French are included.
Brand:  Brady™ 065564
 View more versions of this product
Product Code. 10491064
  • £103.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Our gate valve lockouts have an unique knockout centre accommodates OS and Y or ''rising stem'' valves.
  • Lockouts accept locks with shackles up to 9.53 mm diameter.
  • Danger labels in English, Spanish and French are included.
Specifications
Specifications
Gate Valve Lockout
Red
OSHA 1910.147
13.96 in
354.58 mm
76.20 x 405.10 x 363.50 mm
Lockout Tagout
363.5 mm
15.95 in.
GATE VALVE LOCKOUTS GVLO 10-13 RED
Polypropylene
Brady Gate Valve Lockouts
0.38 in.
9.7 mm
405.10 x 363.50 mm
14.31 in.
1 Ea.
405.1 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Brady™ Polypropylene Gate Valve Lockout > Polypropylene Gate Valve Lockout; Color: Red; Width: 405.1 mm; Height: 363.5 mm