Learn More
Eppendorf™ Polypropylene Graduated Microtubes
Use these tubes for safe handling of valuable and toxic samples as well as radioactive substances. Eppendorf™ Polypropylene Graduated Microtubes feature a hinged lid with a small hook that clips around the rim of the tube.
Brand: Eppendorf™ 0030120191
63.79 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 96
Additional Details : Weight : 1.12770kg
Description
Use these tubes for safe handling of valuable and toxic samples as well as radioactive substances. EppendorfTM Polypropylene Graduated Microtubes feature a hinged lid with a small hook that clips around the rim of the tube.
- Biohazardous substances can be removed, free of aerosols, through a thin membrane in the center of the lid with a cannula
- Lid easily opens with one hand
- These tubes for 0.5mL, 1.5mL and 2mL have a graduation scale and writing surfaces
- The 0.5mL tube is optimized for use in PCR* experiments
- Assorted color packs are a mixture of yellow, red, blue, green and colorless
- Autoclavable micropestle for re-suspending pellets
- Can be centrifuged at up to 23,500xgr
Specifications
30,000 x g | |
1.5 mL | |
Polypropylene | |
1000/Pk. | |
PCR Clean | |
Hinged Snap Cap |
Yes | |
Amber | |
PCR, Centrifugation | |
Sterile | |
Yes |