missing translation for 'onlineSavingsMsg'
Learn More

Ratiolab™ Polypropylene Microcentrifuge Tubes

Use these tubes for a variety of centrifugation applications. Ratiolab™ Polypropylene Microcentrifuge Tubes are available in a variety of colors for easy identification.

Brand:  Ratiolab™ 5615101

 View more versions of this product

Product Code. 10630983

  • £13.30 / Pack of 1000
Estimated Shipment: 13-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Autoclavable with opened cap at 121°C for 20 min.
  • Natural color or different colors for comfortable identification
  • Tight sealing cap
  • Safe and easy to be opened and closed
Specifications

Specifications

Polypropylene
Natural
1000/Pk.
2 mL
Polypropylene
1000/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers