missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ MiniMark™ Polypropylene Unprinted Labels
MiniMark™ Polypropylene Unprinted Labels are ready for you to print the required warning compliant to the latest regulations.
Brand: Brady™ 710680
Code : NEW
Additional Details : Weight : 0.10000kg
Description
- Unprinted labels ready for you to print the required warning compliant to the latest regulations.
Specifications
UL Recognised | |
Permanent Acrylic, Permanent | |
Gloss | |
Polypropylene | |
Labels | |
1 | |
200 Labels/Box | |
Rectangle | |
-4°F to +212°F | |
0.0031 in. | |
160.34 mm | |
6.1 in. | |
4.13 in. |
Unprinted MiniMark Labels for CLP-GHS Hazardous Substances | |
White | |
Label | |
Thermal Transfer | |
Hazard Communication Identification | |
-20°C | |
200 Pk. | |
MiniMark | |
-20°C to +100°C | |
0.079 mm | |
155 x 105 mm | |
155 mm | |
105 mm |