missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ MiniMark™ Polypropylene Unprinted Labels
MiniMark™ Polypropylene Unprinted Labels are ready for you to print the required warning compliant to the latest regulations.
£153.00 - £269.00
Specifications
Certifications/Compliance | UL Recognised |
---|---|
Description | Unprinted MiniMark Labels for CLP-GHS Hazardous Substances |
Adhesive Type | Permanent Acrylic, Permanent |
Color | White |
Finish | Gloss |
Product Code | Brand | Width (Metric) | HeightMetric | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Width (Metric) | HeightMetric | Price | Quantity & Availability | ||||
11787254
|
Brady™
710680 |
105 mm | N/A | ||||||
11797324
|
Brady™
710678 |
75 mm | N/A | ||||||
11707344
|
Brady™
710676 |
55 mm |
£153.00
Each |
Estimated Shipment: 13-01-2025
Log in to see stock. |
|||||
Description
- Unprinted labels ready for you to print the required warning compliant to the latest regulations.
Specifications
UL Recognised | |
Permanent Acrylic, Permanent | |
Gloss | |
Polypropylene | |
Labels | |
-4°F to +212°F | |
0.0031 in. |
Unprinted MiniMark Labels for CLP-GHS Hazardous Substances | |
White | |
Label | |
Thermal Transfer | |
-20°C | |
-20°C to +100°C | |
0.079 mm |