missing translation for 'onlineSavingsMsg'
Learn More

Polypropylene Sterilization Box

Brand:  Azlon PWP050

Product Code. 10787645

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Unique cylindrical box for pipette sterilisation
  • Autoclavable at 121°C
  • The patented telescopic thread closure enables the box length to be altered from 280mm to 500mm
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Polypropylene
Pipette sterilization
Sterilizing boxes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Polypropylene Sterilization Box > Diameter: 65mm; Length: 280 to 500mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.