missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Polypropylene Thread Adapter

Buerkle™ Polypropylene Thread Adapter facilitates the secure connection of pumps and discharge cocks to a variety of containers.

Brand:  Buerkle™ 0600-1007

19.45 GBP valid until 2024-12-31
Use promo code "23267" to get your promotional price.



Additional Details : Weight : 0.20000kg

 View more versions of this product

Product Code. 15953557

  • £20.95 / Each
In Stock
to see stock.
Add to basket

Description

Description

  • Constructed of polypropylene
  • Interchange the connections to different adapters
Specifications

Specifications

Thread Adapter
Polypropylene
External: US drum thread; Internal: 2 in.BSP
Silver
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers