Learn More
Buerkle™ Polypropylene Thread Adapter
Buerkle™ Polypropylene Thread Adapter facilitates the secure connection of pumps and discharge cocks to a variety of containers.
£20.95 - £27.40
Product Code | Brand | Thread Size | Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Thread Size | Color | Price | Quantity & Availability | ||||
10730432
|
Buerkle™
0600-1014 |
External: 64 mm (BSI); Internal: DIN 60 | Black |
£23.15
Each |
In Stock
Log in to see stock. |
||||
10065020
|
Buerkle™
0600-1016 |
External: 64 mm (BSI); Internal: DIN 51 | Black |
£27.40
Each |
In Stock
Log in to see stock. |
||||
10522193
|
Buerkle™
0600-1013 |
External: 64 mm (BSI); Internal: DIN 61 | Yellow, Blue |
£27.40
Each |
In Stock
Log in to see stock. |
||||
10728883
|
Buerkle™
0600-2001 |
Internal: DIN 61 - 2 in. (BSP) | Yellow |
£20.95
Each |
Estimated Shipment: 09-08-2024
Log in to see stock. |
||||
10668283
|
Buerkle™
0600-1006 |
External: Tri-Sure; Internal: 2 in. (BSP) | Orange |
£20.95
Each |
In Stock
Log in to see stock. |
||||
10720763
|
Buerkle™
0600-1001 |
External: 64 mm (BSI); Internal: DIN 71 | Blue, Brown |
£26.55
Each |
In Stock
Log in to see stock. |
||||
11898292
|
Buerkle™
0600-1003 |
Internal: 71 mm; 2 in. (BSP) | Brown |
£20.95
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
||||
15346277
|
Buerkle™
0600-1010 |
External: Mauser; Internal: 2 in. (BSP) | Blue |
£20.95
Each |
In Stock
Log in to see stock. |
||||
15953557
|
Buerkle™
0600-1007 |
External: US drum thread; Internal: 2 in.BSP | Silver |
£20.95
Each |
In Stock
Log in to see stock. |
||||
16358007
|
Buerkle™
0600-2005 |
Internal: 51 mm; 2 in.BSP | Green |
£25.10
Each |
In Stock
Log in to see stock. |
||||
Descrizione
- Constructed of polypropylene
- Interchange the connections to different adapters
Specifica
Thread Adapter | |
Polypropylene |