Learn More
Cole-Parmer™ Essentials Anti-Static Weighing Funnels, Polypropylene
Polypropylene Anti-Static Weighing Funnels ensure complete sample transfer.
Brand: Cole-Parmer™ Essentials 06127-00
Code : NEW
Additional Details : Weight : 0.15000kg
Beschreibung
- Autoclavable
- 6 month warranty
- These disposable polypropylene (PP) weighing funnels are designed to fit in volumetric flasks
- One side of the funnel is flattened so you can set the funnel flat on a balance
Spezifikation
Polypropylene | |
Size: Micro | |
Designed to Fit Microwell Plates |
Autoclavable | |
Volumetric flask | |
Weighing Funnel |