missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polysorbate 80, Vegetable, Multi-Compendial, N.F., J.T. Baker™
£2175.00 - £6362.95
Chemical Identifiers
CAS | 9005-65-6 |
---|---|
Molecular Formula | (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6 |
Molecular Weight (g/mol) | 604.82 |
MDL Number | MFCD00082107 |
InChI Key | HDTIFOGXOGLRCB-KTKRTIGZNA-N |
IUPAC Name | 2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate |
SMILES | CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15508304
|
Avantor J.T.Baker
4500-04 |
200 mL | Narrow Mouth Amber Glass Bottle |
£2175.00
200mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15518304
|
Avantor J.T.Baker
4500-06 |
1 L | Narrow Mouth Amber Glass Bottle | N/A | |||||
Chemical Identifiers
9005-65-6 | |
604.82 | |
HDTIFOGXOGLRCB-KTKRTIGZNA-N | |
CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO |
(C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6 | |
MFCD00082107 | |
2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate |
Specifications
9005-65-6 | |
1L = 1.08kg | |
MFCD00082107 | |
CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO | |
604.82 | |
Polysorbate 80, Vegetable |
148.00°C | |
(C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6 | |
HDTIFOGXOGLRCB-KTKRTIGZNA-N | |
2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate | |
Multi-Compendial/NF |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title