Learn More
Kartell™ Polystyrene Biological Specimen Containers
Manufactured with crystal clear polystyrene for optimal visibility. Kartell™ Polystyrene Biological Specimen Containerrs are available with polyethylene and polypropylene color coded screw caps.
£261.00 - £266.00
Specifications
Product Type Biological Specimen Container
Closure Included Yes
Material Polystyrene
Sterility Sterile
Certifications/Compliance CE and IVD
View More Specs
Products 2
Product Code Brand Capacity (Metric) Diameter (Metric) Height (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Diameter (Metric) Height (Metric) Price Quantity & Availability  
10102381
View Documents Promotion Details
Kartell™
0269600
200 mL 66 mm 86 mm
£261.00
Pack of 200
10355631
View Documents
Kartell™
0563100
60 mL 39 mm 65 mm
£266.00
Pack of 400
Description
Description
  • Sterile containers are individually wrapped
  • CE marked;IVD approved
Specifications
Specifications
Biological Specimen Container
Polystyrene
CE and IVD
Polypropylene
Urine and biological specimen collection
Yes
Yes
Sterile
Red
Screw Cap
Polypropylene Screw Cap