missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heathrow Scientific™ Polystyrene Bulk Wrapped Reagent Reservoir
Reagent Reservoirs offer a fresh and modern design with enhanced features to streamline pipetting procedures.
£69.75
Specifications
Color | White |
---|---|
Material | Polystyrene |
Sterility | Non-sterile |
Type | Reagent Reservoir |
Packaging | Bulk Wrapped |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
16361076
|
Heathrow Scientific™
120752 |
55 mL |
£69.75
Pack of 100 |
Estimated Shipment: 03-01-2025
Log in to see stock. |
|||||
Description
- Trough design with a v shaped bottom helps ensure that all the reagent is accessible and can be easily aspirated
- Thick wall construction and wide base adds stability and durability making the reservoir easy to handle without product deformation which results in less spillage
- Reservoirs can be conveniently stacked with a unique nesting design that enables easy separation
- Designed to stay flat on the benchtop and not warp
- Quality certificate available on request
- Disposable
Specifications
White | |
Non-sterile | |
Bulk Wrapped | |
Not Autoclavable | |
Single-use |
Polystyrene | |
Reagent Reservoir | |
100 Pk. | |
1 | |
Convoluted |