missing translation for 'onlineSavingsMsg'
Learn More

Heathrow Scientific™ Polystyrene Bulk Wrapped Reagent Reservoir

Reagent Reservoirs offer a fresh and modern design with enhanced features to streamline pipetting procedures.

Brand:  Heathrow Scientific™ 120752

59.10 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.


 View more versions of this product

Product Code. 16361076

  • £69.75 / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Trough design with a v shaped bottom helps ensure that all the reagent is accessible and can be easily aspirated
  • Thick wall construction and wide base adds stability and durability making the reservoir easy to handle without product deformation which results in less spillage
  • Reservoirs can be conveniently stacked with a unique nesting design that enables easy separation
  • Designed to stay flat on the benchtop and not warp
  • Quality certificate available on request
  • Disposable
Specifications

Specifications

1.9 oz.
55 mL
Polystyrene
Reagent Reservoir
100 Pk.
1
Convoluted
White
4 x 1.10 x 1.20 in.
Non-sterile
Bulk Wrapped
Not Autoclavable
Single-use
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Heathrow Scientific™ Polystyrene Bulk Wrapped Reagent Reservoir > Capacity: 55 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.