missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heathrow Scientific™ Polystyrene Sterile Single Wrapped Reagent Reservoir
Reagent Reservoirs offer a fresh and modern design with enhanced features to streamline pipetting procedures.
£223.00
Specifications
Color | White |
---|---|
Material | Polystyrene |
Sterility | Sterile |
Type | Reagent Reservoir |
Packaging | Single Wrapped |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
16351076
|
Heathrow Scientific™
120750 |
55 mL |
£223.00
Pack of 100 |
In Stock
Log in to see stock. |
|||||
Description
- Trough design with a v shaped bottom helps ensure that all the reagent is accessible and can be easily aspirated
- Thick wall construction and wide base adds stability and durability making the reservoir easy to handle without product deformation which results in less spillage
- Reservoirs can be conveniently stacked with a unique nesting design that enables easy separation
- Designed to stay flat on the benchtop and not warp
- Sterile, sealed in bags and electron beam sterilized
- DNase, RNase and Pyrogen free
- Sealed in easy-to-open bags
- Quality certificate available on request
- Disposable
Specifications
White | |
Sterile | |
Single Wrapped | |
1 | |
Convoluted |
Polystyrene | |
Reagent Reservoir | |
Not Autoclavable | |
Single-use |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title