missing translation for 'onlineSavingsMsg'
Learn More

Digi-Sense Thermocouple Wire, 24 Gauge

Make your own extension cables with this 24 Gauge Thermocouple Wire.

Brand:  Digi-Sense 08542-12

 View more versions of this product

Product Code. 11705529

  • £329.27 / Each
Estimated Shipment: 15-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Wire comes in 24-gauge size, ideal for fabricating your own probes or extension cables (meets ANSI and ASTM standards).
  • Choose from wire with polyvinyl chloride (PVC), fluorinated ethylene propylene resin (FEP/FEP), fiberglass, high-temperature fiberglass, or ceramic fiber outer/inner insulation.
  • These cables are also available in 100 ft. or 1000 ft. lengths for any application.
Specifications

Specifications

ANSI
Thermocouple Wire
-18.4°F to +429.8°F
300 m
-28°C to +221°C
Polyvinyl
984.3 ft.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers