missing translation for 'onlineSavingsMsg'
Learn More
Polyvinylpyrrolidone, average M.W. 1.300.000, K85-95, Thermo Scientific Chemicals Product Code.: 10125163

Polyvinylpyrrolidone, average M.W. 1.300.000, K85-95, Thermo Scientific Chemicals

Product Code. 10125163
25 g, Plastic bottle
Click to view available options
Quantity:
25 g
500 g
1 kg
2.5 kg
Packaging:
Plastic Drum
Plastic bottle
This item is not returnable. View return policy

Product Code. 10125163

Brand: Thermo Scientific Acros 227470250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9003-39-8
Molecular Formula (C6H9NO)n
Molecular Weight (g/mol) 111.14
MDL Number MFCD01076626
InChI Key WHNWPMSKXPGLAX-UHFFFAOYSA-N
Synonym n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidoneShow MoreShow Less
PubChem CID 6917
ChEBI CHEBI:82551
IUPAC Name 1-ethenylpyrrolidin-2-one
SMILES *-CC(-*)N1CCCC1=O

Specifications

Color Cream-White to White
pH 4 to 7 (5% aq. soln.)
Physical Form Powder
Merck Index 15, 7814
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and chloroform, practically insoluble in ether
Quantity 25 g
Percent Purity 12 to 12.8% (nitrogen)
Identification positive
Infrared Spectrum Authentic
Packaging Plastic bottle
Water 5% max. (K.F.)
Chemical Name or Material Polyvinylpyrrolidone, Average M.W. 1.300.000, K85-95
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468894 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number227470010,227470025,227475000,227470250
Lot NumberA0459306 Certificate TypeCertificate of Analysis Date13/01/2025 Catalog Number227470010,227470025,227475000,227470250
Lot NumberA0450738 Certificate TypeCertificate of Analysis Date26/02/2024 Catalog Number227470010,227470025,227475000,227470250
Lot NumberA0459306 Certificate TypeCertificate of Analysis Date15/01/2024 Catalog Number227470010,227470025,227475000,227470250
Lot NumberA0448628 Certificate TypeCertificate of Analysis Date25/12/2023 Catalog Number227470010,227470025,227475000,227470250
Product Identifier
  • Polyvinylpyrrolidone, average M.W. 1.300.000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Polyvinylpyrrolidone, average M.W. 1.300.000, K85-95, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.