missing translation for 'onlineSavingsMsg'
Learn More
Polyvinylpyrrolidone, M.W. 10,000, Thermo Scientific Chemicals Product Code.: 15423169

Polyvinylpyrrolidone, M.W. 10,000, Thermo Scientific Chemicals

Product Code. 15423169
100 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 15423169

Brand: Thermo Scientific Alfa Aesar J60382.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cryoprotectant, also useful in plant cell culture research

Polyvinylpyrrolidone is a polymer used as a pharmaceutical aid, complexing agent, and solubilizer. PVP was used as a plasma volume expander for trauma victims. It acts as diluent, solvent and cleaning agent in various kinds of industries. It also acts as an adjuvant, emulsifier, filtering aid, tableting aid, thickener.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9003-39-8
Molecular Formula (C6H9NO)n
Molecular Weight (g/mol) 111.14
MDL Number MFCD01076626
InChI Key WHNWPMSKXPGLAX-UHFFFAOYSA-N
Synonym n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone
PubChem CID 6917
ChEBI CHEBI:82551
IUPAC Name 1-ethenylpyrrolidin-2-one
SMILES *-CC(-*)N1CCCC1=O

Specifications

Color White
Physical Form Powder
Merck Index 14,7697
Solubility Information Soluble in water and most organic solvents.
Quantity 100 g
Formula Weight 10 kd
Chemical Name or Material Polyvinylpyrrolidone

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW12J014 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog NumberJ60382.22,J60382.14,J60382.36
Lot NumberY30I031 Certificate TypeCertificate of Analysis Date06/01/2023 Catalog NumberJ60382.22,J60382.14,J60382.36
Lot NumberM24I047 Certificate TypeCertificate of Analysis Date17/11/2022 Catalog NumberJ60382.22,J60382.14,J60382.36
Lot NumberX29H062 Certificate TypeCertificate of Analysis Date17/11/2022 Catalog NumberJ60382.22,J60382.14,J60382.36
Lot NumberY17A013 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberJ60382.22,J60382.14,J60382.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Polyvinylpyrrolidone, M.W. 10,000, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.