missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Pomalidomide Product Code.: 16534504
compliance-icons

Thermo Scientific Chemicals Pomalidomide

Product Code. 16534504
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 16534504

Brand: Thermo Scientific Chemicals 465702500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19171-19-8
Molecular Formula C13H11N3O4
Molecular Weight (g/mol) 273.25
MDL Number MFCD12756407
InChI Key UVSMNLNDYGZFPF-UHFFFAOYNA-N
IUPAC Name 4-amino-2-(2,6-dioxopiperidin-3-yl)-2,3-dihydro-1H-isoindole-1,3-dione
SMILES NC1=CC=CC2=C1C(=O)N(C1CCC(=O)NC1=O)C2=O

Specifications

Melting Point 315.0°C to 320.0°C
Quantity 250 mg
Formula Weight 273.24
Chemical Name or Material Pomalidomide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0453246 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number465702500,465700010
Lot NumberA0415081 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number465702500,465700010
Lot NumberA0447129 Certificate TypeCertificate of Analysis Date03/11/2022 Catalog Number465702500,465700010
Lot NumberA0445204 Certificate TypeCertificate of Analysis Date25/08/2022 Catalog Number465702500,465700010
Lot NumberA0441310 Certificate TypeCertificate of Analysis Date04/05/2022 Catalog Number465702500,465700010
compliance-icons
Product Identifier
  • Pomalidomide
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P403-Store in a well-ventilated place.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Pomalidomide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Pomalidomide >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.