missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ POP-7™ Polymer for Protein Quality Analysis

This is a pouch of POP-7 polymer for protein quality analysis on 3500/3500xL genetic analyzers. The POP-7 separation matrix is validated for protein quality analysis and is offered in easy-to-use pouch packages and 960 sample (maximum) or 384 sample (maximum) size formats.

Brand:  Applied Biosystems™ A31122

590.00 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 15869876

  • £1180.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: Radio frequency identification (RFID) label.
This item is not returnable. View return policy

Description

Description

  • Easy to use: Pre-formulated and in a single-use pouch, which not only saves time but ensures reliability and consistency
  • Robust formulation: Formulated for biomolecular analysis applications
  • Replenishable: Allows capillaries to be used multiple times
  • Includes radio frequency identification (RFID) label: Easily track important information such as part/lot number, samples remaining, and expiry dates (displayed on software dashboard)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at 2–8°C.
3500xL Genetic Analyzer for Protein Quality Analysis
960
Polymer
POP-7™ Polymer For 3500xL Genetic Analyzer
Glycan Analysis, Protein Quality Analysis
POP-7™
Wet Ice
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For research use only