missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Haldenwanger™ Porcelain Evaporating Basins
Use these dishes for a variety of applications. Haldenwanger™ Porcelain Basins feature a smooth internal finish, constructed from optimized laboratory porcelain.
Brand: Haldenwanger™ 131/5
This item is not returnable.
View return policy
Description
Sizes up to 175mL glazed except rim; 1,000mL and larger are glazed except outside base.Specifications
140 mL | |
Round | |
40 mm |
Porcelain | |
Evaporating Basin | |
97 mm |