Learn More
Haldenwanger™ Porcelain Hirsch Funnel
Designed with glazed porcelain for excellent thermal and chemical resistance. Haldenwanger™ Porcelain Hirsch Funnels are an excellent choice for quick filtration.
Brand:  Haldenwanger™ 126/000
 View more versions of this product
Product Code. 10643371
  • £145.00 / Each
Estimated Shipment: 31-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Sloped sides
  • Use for filtering small quantities
  • Perforated fixed filter plate
Specifications
Specifications
52mm
74 mm
15mm
Hirsch Funnel
Porcelain
Size: 000
1Pack