missing translation for 'onlineSavingsMsg'
Learn More

Haldenwanger™ Porcelain Hirsch Funnel

Designed with glazed porcelain for excellent thermal and chemical resistance. Haldenwanger™ Porcelain Hirsch Funnels are an excellent choice for quick filtration.

£145.00 - £155.00

Specifications

Material Porcelain
Quantity 1Pack
Product Type Hirsch Funnel
View More Specs

Products 2
Product Code Brand Description Diameter (Metric) Plate Diameter (Metric)Top Height (Metric) Price Quantity & Availability  
Product Code Brand Description Diameter (Metric) Plate Diameter (Metric)Top Height (Metric) Price Quantity & Availability  
10643371
View Documents
Haldenwanger™
126/000
Size: 000 15mm 52mm 74 mm
£145.00
Each
Estimated Shipment: 27-12-2024
to see stock.
Add to basket
 
10485821
View Documents
Haldenwanger™
126/00
Size: 00 27mm 72mm 102 mm
£155.00
Each
Estimated Shipment: 16-12-2024
to see stock.
Add to basket
 
Description

Description

  • Sloped sides
  • Use for filtering small quantities
  • Perforated fixed filter plate
Specifications

Specifications

Porcelain
1Pack
Hirsch Funnel
Videos
SDS
Documents

Documents

Product Certifications